Thermo Scientific Chemicals

1,9-Dibromononane, 97%, Thermo Scientific Chemicals

Catalog number: 112870250
25 g, Each
Thermo Scientific Chemicals

1,9-Dibromononane, 97%, Thermo Scientific Chemicals

Catalog number: 112870250
25 g, Each
Quantity
Catalog number: 112870250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
4549-33-1
IUPAC Name
1,9-dibromononane
Molecular Formula
C9H18Br2
InChI Key
WGAXVZXBFBHLMC-UHFFFAOYSA-N
SMILES
BrCCCCCCCCCBr
Infrared spectrum
Conforms
GC
>=96.0 %
Appearance (Color)
Clear colorless to yellow
Appearance (Form)
Liquid
Refractive index
1.4950 to 1.4980 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text