Thermo Scientific Chemicals

2,6-Dibromopyridine, 98%, Thermo Scientific Chemicals

Catalog number: 112910250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

2,6-Dibromopyridine, 98%, Thermo Scientific Chemicals

Catalog number: 112910250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 112910250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
626-05-1
IUPAC Name
2,6-dibromopyridine
Molecular Formula
C5H3Br2N
InChI Key
FEYDZHNIIMENOB-UHFFFAOYSA-N
SMILES
BrC1=CC=CC(Br)=N1
Appearance (Color)
White to grey to yellow-brown
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Melting point
117°C to 121°C
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text