alpha,alpha'-Dibromo-o-xylene, 96%, Thermo Scientific Chemicals
alpha,alpha'-Dibromo-o-xylene, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

alpha,alpha'-Dibromo-o-xylene, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Plastic bottle
Catalog number 112931000
Price (USD)/ Each
169.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
199.00 
Save 29.35 (15%)
-
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
169.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
199.00 
Save 29.35 (15%)
Add to cart
alpha,alpha'-Dibromo-o-xylene, 96%, Thermo Scientific Chemicals
Catalog number112931000
Price (USD)/ Each
169.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
199.00 
Save 29.35 (15%)
-
Add to cart
Chemical Identifiers
CAS91-13-4
IUPAC Name1,2-bis(bromomethyl)benzene
Molecular FormulaC8H8Br2
InChI KeyKGKAYWMGPDWLQZ-UHFFFAOYSA-N
SMILESBrCC1=CC=CC=C1CBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to brown
Infrared spectrumConforms
GC>=95 %
Melting point91°C to 96°C
Appearance (Form)Crystalline powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only