alpha,alpha'-Dibromo-p-xylene, 97%, Thermo Scientific Chemicals
alpha,alpha'-Dibromo-p-xylene, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

alpha,alpha'-Dibromo-p-xylene, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number 112941000
Price (USD)/ Each
258.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
304.00 
Save 45.35 (15%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
258.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
304.00 
Save 45.35 (15%)
Add to cart
alpha,alpha'-Dibromo-p-xylene, 97%, Thermo Scientific Chemicals
Catalog number112941000
Price (USD)/ Each
258.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
304.00 
Save 45.35 (15%)
-
Add to cart
Chemical Identifiers
CAS623-24-5
IUPAC Name1,4-bis(bromomethyl)benzene
Molecular FormulaC8H8Br2
InChI KeyRBZMSGOBSOCYHR-UHFFFAOYSA-N
SMILESBrCC1=CC=C(CBr)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Appearance (Form)Crystalline powder or coarse crystals
Infrared spectrumConforms
Melting point142°C to 147°C
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only