Thermo Scientific Chemicals

Di-n-butylamine, 99%, Thermo Scientific Chemicals

Catalog number: 112950100
10 L, Plastic drum, Each
Thermo Scientific Chemicals

Di-n-butylamine, 99%, Thermo Scientific Chemicals

Catalog number: 112950100
10 L, Plastic drum, Each
Quantity
Packaging
Catalog number: 112950100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
111-92-2
IUPAC Name
dibutylamine
Molecular Formula
C8H19N
InChI Key
JQVDAXLFBXTEQA-UHFFFAOYSA-N
SMILES
CCCCNCCCC
Water
=<0.1 %
Refractive index
1.4160 to 1.4180 (20°C, 589 nm)
GC
>=98.5 %
Appearance (Form)
Clear liquid
Color scale
=<20 APHA

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text