Di-n-butylamine, 99%, Thermo Scientific Chemicals
Di-n-butylamine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Di-n-butylamine, 99%, Thermo Scientific Chemicals

Di-n-butylamine, 99%, C8H19N, CAS Number-111-92-2, di-normal-butylamine, n-dibutylamine, n-butyl-1-butanamine, di-n-butyl amine, dibutilamina, dibutylamine, dibutyl amine, di-n-butylamine, dibutyl-amine, 1-butanamine, n-butyl, 2.5L, 98.5% min. (GC), 04, 157, 159.0 deg.C, Undesignated, 0.7600g/mL
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
1 L
2.5 L
10 L
Packaging:
View all
Glass bottle
Catalog number 112950500
Price (USD)/ Each
50.65
Online exclusive
55.60 
Save 4.95 (9%)
-
Add to cart
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
50.65
Online exclusive
55.60 
Save 4.95 (9%)
Add to cart
Di-n-butylamine, 99%, Thermo Scientific Chemicals
Catalog number112950500
Price (USD)/ Each
50.65
Online exclusive
55.60 
Save 4.95 (9%)
-
Add to cart
Chemical Identifiers
CAS111-92-2
IUPAC Namedibutylamine
Molecular FormulaC8H19N
InChI KeyJQVDAXLFBXTEQA-UHFFFAOYSA-N
SMILESCCCCNCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.1 %
Refractive index1.4160 to 1.4180 (20°C, 589 nm)
GC>=98.5 %
Appearance (Form)Clear liquid
Color scale=<20 APHA
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only