Thermo Scientific Chemicals

2,6-Di-tert-butyl-4-methylphenol, 99%, Thermo Scientific Chemicals

Catalog number: 112990050
5 kg, Plastic drum, Each
Thermo Scientific Chemicals

2,6-Di-tert-butyl-4-methylphenol, 99%, Thermo Scientific Chemicals

Catalog number: 112990050
5 kg, Plastic drum, Each
Quantity
Packaging
Catalog number: 112990050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
128-37-0
IUPAC Name
2,6-di-tert-butyl-4-methylphenol
Molecular Formula
C15H24O
InChI Key
NLZUEZXRPGMBCV-UHFFFAOYSA-N
SMILES
CC1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C
Appearance (Color)
Colorless to white
Appearance (Form)
Crystals
Color scale
=<10 APHA (After melting)
Infrared spectrum
Conforms
Melting point
69°C to 71°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text