2,6-Di-tert-butylphenol, 99%, Thermo Scientific Chemicals
2,6-Di-tert-butylphenol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,6-Di-tert-butylphenol, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Packaging:
Glass bottle
Catalog number 113000010
Price (USD)/ Each
-
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS128-39-2
IUPAC Name2,6-di-tert-butylphenol
Molecular FormulaC14H22O
InChI KeyDKCPKDPYUFEZCP-UHFFFAOYSA-N
SMILESCC(C)(C)C1=CC=CC(=C1O)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Crystalline solid
Infrared spectrumConforms
Melting point35°C to 38°C
Water=<0.1 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only