Platinum™ Taq DNA Polymerase, High Fidelity
Platinum&trade; <i>Taq</i> DNA Polymerase, High Fidelity
Invitrogen™

Platinum™ Taq DNA Polymerase, High Fidelity

Platinum Taq DNA Polymerase, High Fidelity, is ideal for amplification of DNA fragments when high yields and robust amplification are required.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
11304011100 Reactions
11304029500 Reactions
113041025000 Reactions
Catalog number 11304011
Price (KRW)
289,000
Online offer
Ends: 30-Jun-2025
321,000
Save 32,000 (10%)
Each
-
Add to cart
No. of Reactions:
100 Reactions
Request bulk or custom format
Price (KRW)
289,000
Online offer
Ends: 30-Jun-2025
321,000
Save 32,000 (10%)
Each
Add to cart
Platinum Taq DNA Polymerase, High Fidelity, is ideal for amplification of DNA fragments when high yields and robust amplification are required. High fidelity is provided by a mixture of Platinum Taq DNA Polymerase and the proofreading enzyme. PCR specificity is improved with built-in Platinum hot-start technology.

Features of Platinum Taq DNA Polymerase, High Fidelity

  • Greater than six times higher fidelity than Taq DNA polymerase
  • Amplification of fragments up to 15 kb
  • Convenient room temperature reaction assembly
  • Increased specificity, and yield

Applications

  • Amplification of complex DNA templates
  • Amplification of viral and plasmid templates
  • RT-PCR
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Fidelity (vs. Taq)6X
Hot StartBuilt-In Hot Start
No. of Reactions100 Reactions
OverhangMixed
PolymerasePlatinum Taq DNA Polymerase High Fidelity
Product TypeDNA Polymerase High Fidelity
Quantity100 rxns
Reaction FormatSeparate Components
Shipping ConditionDry Ice
Size (Final Product)20 kb or less
Detection MethodPrimer-probe
For Use With (Application)Hot-start PCR, High-fidelity PCR
GC-Rich PCR PerformanceLow
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
• Platinum Taq DNA Polymerase High Fidelity (1 x 20 μL at 5 U/μL)
• 10X High Fidelity Buffer (1 x 1.25 mL)
• 50 mM MgSO4 (1 x 1 mL)

Store at -10°C to -30°C.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link