Informational:Inquire about OEM or Commercial Supply version of this product here.
Informational:Upgrade to Invitrogen™ Platinum™ SuperFi™ II DNA Polymerase: >300X higher fidelity than Taq, versatile and now enables universal primer annealing.
Invitrogen™

Platinum™ Taq DNA Polymerase, high fidelity

Catalog number: 11304029
Have Questions?
Invitrogen™

Platinum™ Taq DNA Polymerase, high fidelity

Catalog number: 11304029
Have Questions?
Catalog Number
11304029
Unit Size
500 rxns
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
100 rxns
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
5,000 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
11304029500 rxns
-
11304011100 rxns
-
113041025,000 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

Platinum™ Taq DNA Polymerase, High Fidelity, is ideal for amplification of DNA fragments when high yields and robust amplification are required. High fidelity is provided by a mixture of Platinum™ Taq DNA Polymerase and the proofreading (3´→5´ exonuclease activity) enzyme Pyrococcus species GB-D polymerase. PCR specificity is improved with the incorporation of Platinum™ automatic 'hot-start' technology. Features of Platinum™ Taq DNA Polymerase, High Fidelity:

Fidelity—greater than six times higher fidelity than Taq DNA polymerase
Amplicon size—amplification of fragments up to 15 kb (see figure)
Convenience—room temperature reaction assembly
Applications—amplification of DNA from complex genomic, viral, and plasmid templates; and RT-PCR

Unit definition
One unit of Platinum™ Taq DNA Polymerase, High Fidelity, is the amount of enzyme required to incorporate 10 nmoles of deoxyribonucleotide into DNA in 30 min at 74°C.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
Primer-probe
GC-Rich PCR Performance
Low
Polymerase
Platinum Taq DNA Polymerase High Fidelity
Reaction Speed
Standard
Product Type
DNA Polymerase High Fidelity
Shipping Condition
Dry Ice
For Use With (Application)
Hot-start PCR, High-fidelity PCR
Fidelity (vs. Taq)
6X
Hot Start
Built-In Hot Start
No. of Reactions
500 Reactions
Overhang
Mixed
Reaction Format
Separate Components
Size (Final Product)
20 kb or less

Contents & Storage

• Platinum Taq DNA Polymerase High Fidelity (1 x 100 μL at 5 U/μL)
• 10X High Fidelity Buffer (1 x 2.25 mL)
• 50 mM MgSO4 (1 x 1 mL)

Store at -10°C to -30°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text