1x1 image pixel for data collection
Dichloroacetic acid, 99+%
Dichloroacetic acid, 99+%
Dichloroacetic acid, 99+%
Dichloroacetic acid, 99+%
Thermo Scientific Chemicals

Dichloroacetic acid, 99+%

CAS: 79-43-6 | C2H2Cl2O2 | 128.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
2.5 L
Packaging:
Glass bottle
Catalog number 113060025
Price (USD)
-
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS79-43-6
IUPAC Name2,2-dichloroacetic acid
Molecular FormulaC2H2Cl2O2
InChI KeyJXTHNDFMNIQAHM-UHFFFAOYSA-N
SMILESOC(=O)C(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4650 to 1.4670 (20°C, 589 nm)
Color scale=<100 APHA
Infrared spectrumConforms
Appearance (Form)Clear liquid
Titration with NaOH99.0 to 101.0 %
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0473311Certificate of AnalysisJun 09, 2025113060010, 113060025, 113062500
A0472920Certificate of AnalysisMay 26, 2025113060010, 113060025, 113062500
A0470084Certificate of AnalysisMay 17, 2025113060010, 113060025, 113062500
A0468061Certificate of AnalysisJan 31, 2025113060010, 113060025, 113062500
A0463929Certificate of AnalysisJun 20, 2024113060010, 113060025, 113062500
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection