Thermo Scientific Chemicals

Dichloroacetic acid, 99+%, Thermo Scientific Chemicals

Catalog number: 113062500
250 mL, Glass bottle, Each
Thermo Scientific Chemicals

Dichloroacetic acid, 99+%, Thermo Scientific Chemicals

Catalog number: 113062500
250 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 113062500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
79-43-6
IUPAC Name
2,2-dichloroacetic acid
Molecular Formula
C2H2Cl2O2
InChI Key
JXTHNDFMNIQAHM-UHFFFAOYSA-N
SMILES
OC(=O)C(Cl)Cl
Appearance (Form)
Clear liquid
Color scale
=<100 APHA
Infrared spectrum
Conforms
Refractive index
1.4650 to 1.4670 (20°C, 589 nm)
Titration with NaOH
99.0 to 101.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text