Thermo Scientific Chemicals

2,6-Dichloroaniline, 98%, Thermo Scientific Chemicals

Catalog number: 113100250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

2,6-Dichloroaniline, 98%, Thermo Scientific Chemicals

Catalog number: 113100250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 113100250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
608-31-1
IUPAC Name
2,6-dichloroaniline
Molecular Formula
C6H5Cl2N
InChI Key
JDMFXJULNGEPOI-UHFFFAOYSA-N
SMILES
NC1=C(Cl)C=CC=C1Cl
Appearance (Color)
White to brown
Melting point
35°C to 39°C
GC
>=97.5 %
Appearance (Form)
Crystalline solid
Appearance
May darken on storage

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text