2,6-Dichlorobenzaldehyde, 99%, Thermo Scientific Chemicals
2,6-Dichlorobenzaldehyde, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,6-Dichlorobenzaldehyde, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1 kg
Packaging:
Glass bottle
Catalog number 113162500
Price (USD)/ Each
169.00
-
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
169.00
Add to cart
2,6-Dichlorobenzaldehyde, 99%, Thermo Scientific Chemicals
Catalog number113162500
Price (USD)/ Each
169.00
-
Add to cart
Chemical Identifiers
IUPAC Name2,6-dichlorobenzaldehyde
Molecular FormulaC7H4Cl2O
InChI KeyDMIYKWPEFRFTPY-UHFFFAOYSA-N
SMILESClC1=CC=CC(Cl)=C1C=O
Molecular Weight (g/mol)175.01
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or needles
GC>=98.5 %
Appearance (Color)White to light beige
Infrared spectrumConforms
Melting point68°C to 71°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only