1,2-Dichlorobenzene, 99%, pure, Thermo Scientific Chemicals
1,2-Dichlorobenzene, 99%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,2-Dichlorobenzene, 99%, pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
25 L
Packaging:
View all
Glass Bottle
Catalog number 113180010
Price (USD)/ Each
-
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS95-50-1
IUPAC Name1,2-dichlorobenzene
Molecular FormulaC6H4Cl2
InChI KeyRFFLAFLAYFXFSW-UHFFFAOYSA-N
SMILESClC1=CC=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Appearance (Form)Liquid
Infrared spectrumConforms
Appearance (Color)Clear colorless to light yellow
Refractive index1.5500 to 1.5520 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only