Thermo Scientific Chemicals

1,4-Dichlorobenzene, 99+%, Thermo Scientific Chemicals

Catalog number: 113190025
2.5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

1,4-Dichlorobenzene, 99+%, Thermo Scientific Chemicals

Catalog number: 113190025
2.5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 113190025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
106-46-7
IUPAC Name
1,4-dichlorobenzene
Molecular Formula
C6H4Cl2
InChI Key
OCJBOOLMMGQPQU-UHFFFAOYSA-N
SMILES
ClC1=CC=C(Cl)C=C1
Appearance (Color)
White
Appearance (Form)
Crystals
Melting point
52°C to 55°C
Infrared spectrum
Conforms
GC
>=99.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text