Thermo Scientific Chemicals

2,6-Dichlorobenzoic Acid, 98+%, Thermo Scientific Chemicals

Catalog number: 113200100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

2,6-Dichlorobenzoic Acid, 98+%, Thermo Scientific Chemicals

Catalog number: 113200100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 113200100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
50-30-6
IUPAC Name
2,6-dichlorobenzoic acid
Molecular Formula
C7H4Cl2O2
InChI Key
MRUDNSFOFOQZDA-UHFFFAOYSA-N
SMILES
OC(=O)C1=C(Cl)C=CC=C1Cl
Appearance (Color)
White to beige
Infrared spectrum
Conforms
HPLC
>=98.0 %
Appearance (Form)
Crystalline powder
Melting point
141°C to 145°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text