Thermo Scientific Chemicals

2,3-Dichloro-5,6-dicyano-1,4-benzoquinone, 98%, Thermo Scientific Chemicals

Catalog number: 113300100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

2,3-Dichloro-5,6-dicyano-1,4-benzoquinone, 98%, Thermo Scientific Chemicals

Catalog number: 113300100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 113300100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
84-58-2
IUPAC Name
4,5-dichloro-3,6-dioxocyclohexa-1,4-diene-1,2-dicarbonitrile
Molecular Formula
C8Cl2N2O2
InChI Key
HZNVUJQVZSTENZ-UHFFFAOYSA-N
SMILES
ClC1=C(Cl)C(=O)C(C#N)=C(C#N)C1=O
Infrared spectrum
Conforms
Appearance (Color)
Yellow to orange
Melting point
210°C to 217°C
Loss on drying
=<0.5 % (1 g, 80°C, 2 h, 40 mmHg) (vacuum)
Appearance (Form)
Crystalline powder or crystals

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text