DMEM/F-12, HEPES
Save up to 29% on your order of cell culture products. View promotion details >
DMEM/F-12, HEPES
Gibco™

DMEM/F-12, HEPES

Dulbecco's Modified Eagle Medium:Nutrient Mixture F-12 (DMEM / F-12) is a widely used basal medium for supporting the growth ofRead more
Promo ImagePromotion
Promo code:P5906365
Don't miss out: buy 3, only pay for 2
Promo ImagePromotion
Promo code:RPUZZ25
Stock up on essentials to piece your discovery together
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
11330057Promo Image10 x 500 mL
11330032Promo Image500 mL
11330099Promo Image5 L
11330107Promo Image10 L
Catalog number 11330057
Price (USD)
419.65
Online Exclusive
459.00
Save 39.35 (9%)
Each
Add to cart
Quantity:
10 x 500 mL
Customize this product
Price (USD)
419.65
Online Exclusive
459.00
Save 39.35 (9%)
Each
Add to cart

Dulbecco's Modified Eagle Medium:Nutrient Mixture F-12 (DMEM / F-12) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM / F-12 include MDCK, glial cells, fibroblasts, human endothelial cells, and rat fibroblasts. We offer a variety of Gibco™ DMEM / F-12 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This DMEM / F-12 is modified as follows:

With: L-glutamine, HEPES, Phenol Red

The complete formulation is available.Gibco™ DMEM / F-12 is a 1:1 mixture of DMEM and Ham's F-12. This formulation combines DMEM's high concentrations of glucose, amino acids, and vitamins with F-12's wide variety of components.

DMEM / F-12 contains no proteins, lipids, or growth factors. Therefore, DMEM / F-12 may require supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM / F-12 uses a sodium bicarbonate buffer system and therefore requires a 5-10% CO2 environment to maintain physiological pH.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Cell LineMDCK, glial cells, fibroblasts, human endothelial cells, and rat fibroblasts
Concentration1 X
For Use With (Application)Mammalian cell culture
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Product LineGibco™
Product TypeDMEM (Dulbecco's Modified Eagle Medium)/F-12
Quantity10 x 500 mL
Shelf Life12 Months From Date of Manufacture
ClassificationAnimal Origin-free
FormLiquid
Serum LevelStandard Serum Supplementation
SterilitySterile-filtered
Sterilization MethodSterile-filtered
With AdditivesHigh Glucose, Glutamine, HEPES, Phenol Red, Sodium Pyruvate
Unit SizeEach
Contents & Storage
Storage conditions: 2°C to 8°C (protect from light)
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Media Formulations

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link