2,3-Dichloro-5,6-dicyano-1,4-benzoquinone, 98%, Thermo Scientific Chemicals
2,3-Dichloro-5,6-dicyano-1,4-benzoquinone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,3-Dichloro-5,6-dicyano-1,4-benzoquinone, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
10 g
100 g
Packaging:
Glass bottle
Catalog number 113305000
Price (USD)/ Each
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS84-58-2
IUPAC Name4,5-dichloro-3,6-dioxocyclohexa-1,4-diene-1,2-dicarbonitrile
Molecular FormulaC8Cl2N2O2
InChI KeyHZNVUJQVZSTENZ-UHFFFAOYSA-N
SMILESClC1=C(Cl)C(=O)C(C#N)=C(C#N)C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Yellow to orange
Melting point210°C to 217°C
Loss on drying=<0.5 % (1 g, 80°C, 2 h, 40 mmHg) (vacuum)
Appearance (Form)Crystalline powder or crystals
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only