Dynabeads™ CD4 Positive Isolation Kit
Dynabeads™ CD4 Positive Isolation Kit
Invitrogen™

Dynabeads™ CD4 Positive Isolation Kit

The Dynabeads™ CD4 Positive Isolation Kit supplies superparamagnetic beads that enable gentle isolation of high-purity CD4+ T cells from wholeRead more
Have Questions?
Catalog NumberQuantity
11331D5 mL
Catalog number 11331D
Price (EUR)
1.390,00
5 mL
-
Add to cart
Quantity:
5 mL
Price (EUR)
1.390,00
5 mL
Add to cart
Ask our AI about this Product
The Dynabeads™ CD4 Positive Isolation Kit supplies superparamagnetic beads that enable gentle isolation of high-purity CD4+ T cells from whole blood, bone marrow, buffy coat, mononuclear cells (MNC), or tissue digests. The cells are released from the beads using the DETACHaBEAD™ reagent to obtain bead- and antibody-free cells. Advantages of the Dynabeads™ CD4 Positive Isolation Kit:

• Gentle isolation of CD4+ T cells from any sample—no columns required
• Bead- and antibody-free positive isolation of CD4+ cells for any downstream assay

About Dynabeads™ CD4 Positive Isolation Kit
The Dynabeads™ CD4 Positive Isolation Kit contains uniform, superparamagnetic beads (4.5 μm diameter) covalently coated with a monoclonal antibody specific for the CD4 membrane antigen predominantly expressed on the helper/ inducer subset of human T cells. The kit also includes a DETACHaBEAD™ reagent that releases the Dynabeads™ from the purified CD4+ cells. This gently magnetic isolation process gives highly viable bead- and antibody-free cells to be used in any downstream assay.

Magnetic bead-based separation offers easy handling
Dynabeads™ are mixed with the sample in a tube where they bind to the target cells during a short incubation, after which the bead-bound cells are separated from unbound cells using a magnet. The positively isolated cells are then washed, and DETACHaBEAD™ reagent is added to gently release the cells from the beads. This magnetic bead-based positive isolation method gives both bead- and antibody-free cells. The isolated CD4+ cells are phenotypically unaltered and suitable for any downstream application. This fast and gentle isolation method does not require the use of columns, and helps ensure high purity and viability of the isolated CD4+ cells. For depletion or isolation of untouched CD4+ cells, select from our range of Human CD4+ T Cell Isolation Products.

Learn more about Dynabeads™ products
• Find Dynabeads™ products for a whole range of applications.
• Find magnets for Dynabeads™ separations.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeT Cells (CD4)
Isolation TechnologyPositive Isolation
No. of CellsProcesses ∼2 x 109 cells total
Output Viability>95%
Product LineDETACHaBEAD™, DYNAL™, Dynabeads™
Purity or Quality GradeResearch Grade
Quantity5 mL
Sample TypePBMC, Blood
Shipping ConditionRoom Temperature
Starting Material Cell No.1 x 107 PBMCs per isolation
Target SpeciesHuman
Diameter (Metric)4.5 μm
Product TypePositive Cell Isolation Kit
Unit Size5 mL
Contents & Storage
Each kit contains 5 mL Dynabeads™ CD4 coated with anti-CD4 monoclonal antibody, and 2 mL DETACHaBEAD™ CD4. The kit can process up to 2 x 109 cells.

Storage Conditions: 2°C to 8°C. Do not freeze.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link