5,7-Dichloro-8-hydroxyquinoline, 98%, Thermo Scientific Chemicals
5,7-Dichloro-8-hydroxyquinoline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5,7-Dichloro-8-hydroxyquinoline, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
Packaging:
Glass bottle
Catalog number 113431000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS773-76-2
IUPAC Name5,7-dichloroquinolin-8-ol
Molecular FormulaC9H5Cl2NO
InChI KeyWDFKMLRRRCGAKS-UHFFFAOYSA-N
SMILESOC1=C2N=CC=CC2=C(Cl)C=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Appearance (Color)White to beige to grey
Infrared spectrumConforms
Appearance (Form)Powder
Melting point178°C to 183°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only