Gibco™

HEPES (Ultra Pure)

Catalog number: 11344041
Have Questions?
Gibco™

HEPES (Ultra Pure)

Catalog number: 11344041
Have Questions?
Catalog Number
11344041
Unit Size
1 kg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
113440411 kg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Gibco™ HEPES (N-2-Hydroxyethylpiperazine-N-2-Ethane Sulfonic Acid) is a zwitterionic organic chemical buffering agent commonly used in cell culture media. The addition of 10 – 25 mM HEPES provides extra buffering capacity when cell culture requires extended periods of manipulation outside of a CO2 incubator. HEPES is a good buffering choice for many cell culture systems because it is membrane impermeable, has limited effect on biochemical reactions, is chemically and enzymatically stable, and has very low visible and UV light absorbance.

Product Use
For Research Use Only. Not for use in diagnostic procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Type
Mammalian Cells
Form
Powder
Product Type
HEPES Buffer
Shelf Life
24 Months
pH
6.8 to 8.2
Shipping Condition
Room Temperature

Contents & Storage

Storage conditions: 15 – 30°C
Shipping conditions: Ambient
Shelf life: 24 months from date of manufacture

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text