Thermo Scientific Chemicals

2,3-Dichloromaleic anhydride, 97%, Thermo Scientific Chemicals

Catalog number: 113441000
100 g, Each
Thermo Scientific Chemicals

2,3-Dichloromaleic anhydride, 97%, Thermo Scientific Chemicals

Catalog number: 113441000
100 g, Each
Quantity
Catalog number: 113441000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1122-17-4
IUPAC Name
dichloro-2,5-dihydrofuran-2,5-dione
Molecular Formula
C4Cl2O3
InChI Key
AGULWIQIYWWFBJ-UHFFFAOYSA-N
SMILES
ClC1=C(Cl)C(=O)OC1=O
Appearance (Color)
White to yellow
Appearance (Form)
Crystalline powder or solid
Infrared spectrum
Conforms
GC
>=96 %
NMR (1H-NMR)
Consistent with structure

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text