Thermo Scientific Chemicals

1,1-Dichlorodimethyl ether, 97%, Thermo Scientific Chemicals

Catalog number: 113471000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

1,1-Dichlorodimethyl ether, 97%, Thermo Scientific Chemicals

Catalog number: 113471000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 113471000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4885-02-3
IUPAC Name
dichloro(methoxy)methane
Molecular Formula
C2H4Cl2O
InChI Key
GRTGGSXWHGKRSB-UHFFFAOYSA-N
SMILES
COC(Cl)Cl
Appearance (Color)
Clear colorless to light yellow
Refractive index
1.4295 to 1.4355 (20°C, 589 nm)
Infrared spectrum
Conforms
GC
>=96.0 %
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text