Thermo Scientific Chemicals

2,3-Dichloro-1,4-naphthoquinone, 98%, Thermo Scientific Chemicals

Catalog number: 113481000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

2,3-Dichloro-1,4-naphthoquinone, 98%, Thermo Scientific Chemicals

Catalog number: 113481000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 113481000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
117-80-6
IUPAC Name
2,3-dichloro-1,4-dihydronaphthalene-1,4-dione
Molecular Formula
C10H4Cl2O2
InChI Key
SVPKNMBRVBMTLB-UHFFFAOYSA-N
SMILES
ClC1=C(Cl)C(=O)C2=CC=CC=C2C1=O
Appearance (Form)
Crystalline powder
GC
>=97.5 %
Appearance (Color)
Yellow
Melting point
194°C to 197°C
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text