2,5-Dichlorophenol, 98%, Thermo Scientific Chemicals
2,5-Dichlorophenol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,5-Dichlorophenol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1 kg
Packaging:
Glass bottle
Catalog number 113582500
Price (USD)/ Each
150.00
-
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
150.00
Add to cart
2,5-Dichlorophenol, 98%, Thermo Scientific Chemicals
Catalog number113582500
Price (USD)/ Each
150.00
-
Add to cart
Chemical Identifiers
CAS583-78-8
IUPAC Name2,5-dichlorophenol
Molecular FormulaC6H4Cl2O
InChI KeyRANCECPPZPIPNO-UHFFFAOYSA-N
SMILESOC1=CC(Cl)=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals of flakes
Melting point54°C to 58°C
Appearance (Color)White to light brown
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only