2,6-Dichlorophenol, 99%, Thermo Scientific Chemicals
2,6-Dichlorophenol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,6-Dichlorophenol, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 113590250
Price (USD)/ Each
26.65
Online exclusive
30.70 
Save 4.05 (13%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
26.65
Online exclusive
30.70 
Save 4.05 (13%)
Add to cart
2,6-Dichlorophenol, 99%, Thermo Scientific Chemicals
Catalog number113590250
Price (USD)/ Each
26.65
Online exclusive
30.70 
Save 4.05 (13%)
-
Add to cart
Chemical Identifiers
IUPAC Name2,6-dichlorophenol
Molecular FormulaC6H4Cl2O
InChI KeyHOLHYSJJBXSLMV-UHFFFAOYSA-N
SMILESOC1=C(Cl)C=CC=C1Cl
Molecular Weight (g/mol)163.00
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Melting point65°C to 68°C
Infrared spectrumConforms
Appearance (Form)Crystalline powder or crystals
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only