Thermo Scientific Chemicals

2,6-Dichlorophenol, 99%, Thermo Scientific Chemicals

Catalog number: 113591000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

2,6-Dichlorophenol, 99%, Thermo Scientific Chemicals

Catalog number: 113591000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 113591000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
87-65-0
IUPAC Name
2,6-dichlorophenol
Molecular Formula
C6H4Cl2O
InChI Key
HOLHYSJJBXSLMV-UHFFFAOYSA-N
SMILES
OC1=C(Cl)C=CC=C1Cl
Appearance (Color)
White to yellow
Melting point
65°C to 68°C
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder or crystals
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text