1,3-Dichloropropane, 99%, Thermo Scientific Chemicals
1,3-Dichloropropane, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,3-Dichloropropane, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
50 mL
Packaging:
Glass bottle
Catalog number 113682500
Price (USD)/ Each
-
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS142-28-9
IUPAC Name1,3-dichloropropane
Molecular FormulaC3H6Cl2
InChI KeyYHRUOJUYPBUZOS-UHFFFAOYSA-N
SMILESClCCCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless
Infrared spectrumConforms
Refractive index1.4471 to 1.4491 (20°C, 589 nm)
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only