Thermo Scientific Chemicals

2,3-Dichloro-1-propene, 98%, Thermo Scientific Chemicals

Catalog number: 113691000
100 mL, Glass bottle, Each
Thermo Scientific Chemicals

2,3-Dichloro-1-propene, 98%, Thermo Scientific Chemicals

Catalog number: 113691000
100 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 113691000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
78-88-6
IUPAC Name
2,3-dichloroprop-1-ene
Molecular Formula
C3H4Cl2
InChI Key
FALCMQXTWHPRIH-UHFFFAOYSA-N
SMILES
ClCC(Cl)=C
GC
>=97.5 %
Appearance (Color)
Clear colorless to pink-brown
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Refractive index
1.4601 to 1.4621 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text