Thermo Scientific Chemicals

2,3-Dichloropyridine, 99%, Thermo Scientific Chemicals

Catalog number: 113720100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

2,3-Dichloropyridine, 99%, Thermo Scientific Chemicals

Catalog number: 113720100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 113720100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2402-77-9
IUPAC Name
2,3-dichloropyridine
Molecular Formula
C5H3Cl2N
InChI Key
MAKFMOSBBNKPMS-UHFFFAOYSA-N
SMILES
ClC1=CC=CN=C1Cl
Melting point
67°C to 70°C
Appearance (Color)
White to light beige
Infrared spectrum
Conforms
GC
>=98.5 %
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text