Thermo Scientific Chemicals

3,5-Dichloropyridine, 97%, Thermo Scientific Chemicals

Catalog number: 113741000
Each
Thermo Scientific Chemicals

3,5-Dichloropyridine, 97%, Thermo Scientific Chemicals

Catalog number: 113741000
Each
Catalog number: 113741000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2457-47-8
Appearance (Form)
Powder
Melting point
64°C to 68°C
Appearance (Color)
White to yellow
Infrared spectrum
Conforms
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text