Thermo Scientific Chemicals

Dicyandiamide, 99.5%, Thermo Scientific Chemicals

Catalog number: 113810050
5 g, Glass Bottle, Each
Thermo Scientific Chemicals

Dicyandiamide, 99.5%, Thermo Scientific Chemicals

Catalog number: 113810050
5 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 113810050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
461-58-5
IUPAC Name
N''-cyanoguanidine
Molecular Formula
C2H4N4
InChI Key
QGBSISYHAICWAH-UHFFFAOYSA-N
SMILES
NC(N)=NC#N
Sulfated ash
=<0.1 %
Appearance (Color)
White
Infrared spectrum
Conforms
Melting point
208°C to 211°C
Water
=<0.5 % (K.F.)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text