Dicyandiamide, 99.5%
Dicyandiamide, 99.5%
Dicyandiamide, 99.5%
Dicyandiamide, 99.5%
Thermo Scientific Chemicals

Dicyandiamide, 99.5%

CAS: 461-58-5 | C2H4N4 | 84.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
1 kg
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number 113810250
Price (USD)
18.65
Special offer
Online exclusive
Ends: 26-Sep-2025
23.10
Save 4.45 (19%)
Each
-
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
18.65
Special offer
Online exclusive
Ends: 26-Sep-2025
23.10
Save 4.45 (19%)
Each
Add to cart
Chemical Identifiers
CAS461-58-5
IUPAC NameN''-cyanoguanidine
Molecular FormulaC2H4N4
InChI KeyQGBSISYHAICWAH-UHFFFAOYSA-N
SMILESNC(N)=NC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Sulfated ash=<0.1 %
Appearance (Color)White
Infrared spectrumConforms
Melting point208°C to 211°C
Water=<0.5 % (K.F.)
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0469222Certificate of AnalysisJan 31, 2025113810250, 113810010, 113810025, 113810050
A0463238Certificate of AnalysisMay 31, 2024113810250, 113810010, 113810025, 113810050
A0393187Certificate of AnalysisDec 15, 2022113810250, 113810010, 113810025, 113810050
A0409305Certificate of AnalysisDec 13, 2022113810250, 113810010, 113810025, 113810050
A0406750Certificate of AnalysisDec 13, 2022113810250, 113810010, 113810025, 113810050
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection