Thermo Scientific Chemicals

1,4-Dicyanobenzene, 98%, Thermo Scientific Chemicals

Catalog number: 113821000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

1,4-Dicyanobenzene, 98%, Thermo Scientific Chemicals

Catalog number: 113821000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 113821000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
623-26-7
IUPAC Name
benzene-1,4-dicarbonitrile
Molecular Formula
C8H4N2
InChI Key
BHXFKXOIODIUJO-UHFFFAOYSA-N
SMILES
N#CC1=CC=C(C=C1)C#N
Appearance (Color)
White to light yellow to light beige
Appearance (Form)
Powder
Infrared spectrum
Conforms
Melting point
223°C to 230°C
HPLC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text