Thermo Scientific Chemicals

1,4-Dicyanobutane, 99%, Thermo Scientific Chemicals

Catalog number: 113830010
1 L, Glass bottle, Each
Thermo Scientific Chemicals

1,4-Dicyanobutane, 99%, Thermo Scientific Chemicals

Catalog number: 113830010
1 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 113830010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
111-69-3
IUPAC Name
hexanedinitrile
Molecular Formula
C6H8N2
InChI Key
BTGRAWJCKBQKAO-UHFFFAOYSA-N
SMILES
N#CCCCCC#N
Infrared spectrum
Conforms
Refractive index
1.4370 to 1.4390 (20°C, 589 nm)
Appearance (Form)
Liquid
Appearance (Color)
Clear colorless to light yellow to light orange
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text