N,N'-Dicyclohexylcarbodiimide, 99%
N,N'-Dicyclohexylcarbodiimide, 99%
N,N'-Dicyclohexylcarbodiimide, 99%
N,N'-Dicyclohexylcarbodiimide, 99%
Thermo Scientific Chemicals

N,N'-Dicyclohexylcarbodiimide, 99%

N,N-Dicyclohexylcarbodiimide, 99%, C13H22N2, CAS Number-538-75-0, carbodiimide, dicyclohexyl, bis cyclohexyl carbodiimide, dcc, 1,3-dicyclohexylcarbodiimide, carbodicyclohexylimide, n,n'-dicyclohexylcarbodiimide, n,n-dicyclohexylcarbodiimide, dcci, dicyclohexylcarbodiimide, dccd, 1kg | CAS: 538-75-0 | C13H22N2 | 206.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
100 g
250 g
5 kg
25 kg
Packaging:
View all
Glass bottle
Catalog number 113900010
Price (USD)
272.00
Each
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
272.00
Each
Add to cart
Chemical Identifiers
CAS538-75-0
IUPAC NameN,N'-dicyclohexylmethanediimine
Molecular FormulaC13H22N2
InChI KeyQOSSAOTZNIDXMA-UHFFFAOYSA-N
SMILESC1CCC(CC1)N=C=NC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Crystalline mass or waxy solid
Infrared spectrumConforms
Melting point33°C to 35°C
GC>=98.5 %
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Kunden, die diesen Artikel ansahen, interessierten sich auch für



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0442027Certificate of AnalysisMay 19, 2025113900010, 113900250, 113901000, 113900050, 113902500
A0447018Certificate of AnalysisApr 29, 2024113900010, 113900250, 113901000, 113900050, 113902500
A0442027Certificate of AnalysisNov 17, 2023113900010, 113900250, 113901000, 113900050, 113902500
A0432446Certificate of AnalysisJan 19, 2023113900010, 113900250, 113901000, 113900050, 113902500
A0429091Certificate of AnalysisJan 17, 2023113900010, 113900250, 113901000, 113900050, 113902500
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection