Thermo Scientific Chemicals

N,N'-Dicyclohexylcarbodiimide, 99%, Thermo Scientific Chemicals

Catalog number: 113900250
25 kg, Plastic drum, Each
Thermo Scientific Chemicals

N,N'-Dicyclohexylcarbodiimide, 99%, Thermo Scientific Chemicals

Catalog number: 113900250
25 kg, Plastic drum, Each
Quantity
Packaging
Catalog number: 113900250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
538-75-0
IUPAC Name
N,N'-dicyclohexylmethanediimine
Molecular Formula
C13H22N2
InChI Key
QOSSAOTZNIDXMA-UHFFFAOYSA-N
SMILES
C1CCC(CC1)N=C=NC1CCCCC1
Appearance (Color)
White to light yellow
Appearance (Form)
Crystalline mass or waxy solid
Infrared spectrum
Conforms
Melting point
33°C to 35°C
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text