Thermo Scientific Chemicals

N,N'-Dicyclohexylurea, 98%, Thermo Scientific Chemicals

Catalog number: 113910250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

N,N'-Dicyclohexylurea, 98%, Thermo Scientific Chemicals

Catalog number: 113910250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 113910250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2387-23-7
IUPAC Name
1,3-dicyclohexylurea
Molecular Formula
C13H24N2O
InChI Key
ADFXKUOMJKEIND-UHFFFAOYSA-N
SMILES
O=C(NC1CCCCC1)NC1CCCCC1
Appearance (Color)
White to light yellow
Titration with HClO4
>=97.5 %
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder
Melting point
231°C to 236°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text