Thermo Scientific Chemicals

Diethanolamine, 99%, Thermo Scientific Chemicals

Catalog number: 113920010
1 kg, Glass Bottle, Each
Thermo Scientific Chemicals

Diethanolamine, 99%, Thermo Scientific Chemicals

Catalog number: 113920010
1 kg, Glass Bottle, Each
Quantity
Packaging
Catalog number: 113920010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
111-42-2
IUPAC Name
2-[(2-hydroxyethyl)amino]ethan-1-ol
Molecular Formula
C4H11NO2
InChI Key
ZBCBWPMODOFKDW-UHFFFAOYSA-N
SMILES
OCCNCCO
Infrared spectrum
Conforms
Water
=<0.2 % (K.F.)
Color scale
=<20 APHA
Refractive index
1.4760 to 1.4790 (20°C, 589 nm)
Appearance (Form)
Clear viscous liquid or low melting solid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text