Diethanolamine, 99%, Thermo Scientific Chemicals
Diethanolamine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diethanolamine, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
1 kg
25 kg
Packaging:
View all
Glass Bottle
Catalog number 113920025
Price (USD)/ Each
103.65
Online exclusive
115.00 
Save 11.35 (10%)
-
Add to cart
Quantity:
2.5 kg
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
103.65
Online exclusive
115.00 
Save 11.35 (10%)
Add to cart
Diethanolamine, 99%, Thermo Scientific Chemicals
Catalog number113920025
Price (USD)/ Each
103.65
Online exclusive
115.00 
Save 11.35 (10%)
-
Add to cart
Chemical Identifiers
CAS111-42-2
IUPAC Name2-[(2-hydroxyethyl)amino]ethan-1-ol
Molecular FormulaC4H11NO2
InChI KeyZBCBWPMODOFKDW-UHFFFAOYSA-N
SMILESOCCNCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Water=<0.2 % (K.F.)
Color scale=<20 APHA
Refractive index1.4760 to 1.4790 (20°C, 589 nm)
Appearance (Form)Clear viscous liquid or low melting solid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only