Thermo Scientific Chemicals

Diethyl Acetamidomalonate, 99+%, Thermo Scientific Chemicals

Catalog number: 113930250
25 g, Plastic bottle, Each
Thermo Scientific Chemicals

Diethyl Acetamidomalonate, 99+%, Thermo Scientific Chemicals

Catalog number: 113930250
25 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 113930250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1068-90-2
IUPAC Name
1,3-diethyl 2-acetamidopropanedioate
Molecular Formula
C9H15NO5
InChI Key
ISOLMABRZPQKOV-UHFFFAOYSA-N
SMILES
CCOC(=O)C(NC(C)=O)C(=O)OCC
Appearance (Color)
White to light yellow
Infrared spectrum
Conforms
GC
>=99.0 %
Appearance (Form)
Crystalline powder
Melting point
94°C to 97°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text