Thermo Scientific Chemicals

Diethyl acetylsuccinate, 99%, Thermo Scientific Chemicals

Catalog number: 113950250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Diethyl acetylsuccinate, 99%, Thermo Scientific Chemicals

Catalog number: 113950250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 113950250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1115-30-6
IUPAC Name
1,4-diethyl 2-acetylbutanedioate
Molecular Formula
C10H16O5
InChI Key
DVSDDICSXBCMQJ-UHFFFAOYNA-N
SMILES
CCOC(=O)CC(C(C)=O)C(=O)OCC
Appearance (Color)
Clear colorless to light yellow
GC
>=98.5 %
Infrared spectrum
Conforms
Appearance (Form)
Liquid
Refractive index
1.4340 to 1.4360 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text