Thermo Scientific Chemicals

4-(Diethylamino)benzaldehyde, 99%, Thermo Scientific Chemicals

Catalog number: 114005000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

4-(Diethylamino)benzaldehyde, 99%, Thermo Scientific Chemicals

Catalog number: 114005000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 114005000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
120-21-8
IUPAC Name
4-(diethylamino)benzaldehyde
Molecular Formula
C11H15NO
InChI Key
MNFZZNNFORDXSV-UHFFFAOYSA-N
SMILES
CCN(CC)C1=CC=C(C=O)C=C1
Infrared spectrum
Conforms
Titration with HClO4
>=98.5 %
Appearance (Form)
Coarse crystals or chunks or crystalline solid
Melting point
37°C to 41°C
Appearance (Color)
Green to brown to black

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text