2-Diethylaminoethylchloride hydrochloride, 99.5%, Thermo Scientific Chemicals
2-Diethylaminoethylchloride hydrochloride, 99.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Diethylaminoethylchloride hydrochloride, 99.5%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 g
250 g
2.5 kg
Packaging:
View all
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 114030010
Price (USD)/ Each
-
Quantity:
1 kg
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namehydrogen (2-chloroethyl)diethylamine chloride
Molecular FormulaC6H15Cl2N
InChI KeyRAGSWDIQBBZLLL-UHFFFAOYSA-N
SMILES[H+].[Cl-].CCN(CC)CCCl
Molecular Weight (g/mol)172.09
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with HClO4>=99.4 % (On dry substance)
Appearance (Form)Crystalline powder
Melting point208°C to 212°C
Loss on drying=<1 % (105°C, 3 hrs)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only