Thermo Scientific Chemicals

2-Diethylaminoethylchloride hydrochloride, 99.5%, Thermo Scientific Chemicals

Catalog number: 114032500
250 g, Glass Bottle, Each
Thermo Scientific Chemicals

2-Diethylaminoethylchloride hydrochloride, 99.5%, Thermo Scientific Chemicals

Catalog number: 114032500
250 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 114032500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
869-24-9
IUPAC Name
hydrogen (2-chloroethyl)diethylamine chloride
Molecular Formula
C6H15Cl2N
InChI Key
RAGSWDIQBBZLLL-UHFFFAOYSA-N
SMILES
[H+].[Cl-].CCN(CC)CCCl
Titration with HClO4
>=99.4 % (On dry substance)
Appearance (Form)
Crystalline powder
Melting point
208°C to 212°C
Loss on drying
=<1 % (105°C, 3 hrs)
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text