Thermo Scientific Chemicals

Diethyl aminomalonate hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 114040250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Diethyl aminomalonate hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 114040250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 114040250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
13433-00-6
IUPAC Name
hydrogen 1,3-diethyl 2-aminopropanedioate chloride
Molecular Formula
C7H14ClNO4
InChI Key
GLFVNTDRBTZJIY-UHFFFAOYSA-N
SMILES
[H+].[Cl-].CCOC(=O)C(N)C(=O)OCC
Appearance (Form)
Crystalline powder
Appearance (Color)
White to light yellow
Infrared spectrum
Conforms
Titration Argentometric
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text