Thermo Scientific Chemicals

7-Diethylamino-4-methylcoumarin, 99%

Catalog number: 114055000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

7-Diethylamino-4-methylcoumarin, 99%

Catalog number: 114055000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 114055000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
91-44-1
IUPAC Name
7-(diethylamino)-4-methyl-2H-chromen-2-one
Molecular Formula
C14H17NO2
InChI Key
AFYCEAFSNDLKSX-UHFFFAOYSA-N
SMILES
CCN(CC)C1=CC=C2C(C)=CC(=O)OC2=C1
Appearance (Color)
Light beige to light purple
Infrared spectrum
Conforms
GC
>=98.5 %
Appearance (Form)
Crystalline powder
Melting point
70°C to 73°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text