3-Diethylaminopropylamine, 99+%, Thermo Scientific Chemicals
3-Diethylaminopropylamine, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Diethylaminopropylamine, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 114092500
Price (USD)/ Each
44.10
-
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
44.10
Add to cart
3-Diethylaminopropylamine, 99+%, Thermo Scientific Chemicals
Catalog number114092500
Price (USD)/ Each
44.10
-
Add to cart
Chemical Identifiers
CAS104-78-9
IUPAC Name(3-aminopropyl)diethylamine
Molecular FormulaC7H18N2
InChI KeyQOHMWDJIBGVPIF-UHFFFAOYSA-N
SMILESCCN(CC)CCCN
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.0 %
Refractive index1.4406 to 1.4426 (20°C, 589 nm)
Appearance (Form)Liquid
Water=<0.3 %
Appearance (Color)Clear colorless to light yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only